Methyl 2-(difluoromethyl)pyrazole-3-carboxylate 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2101195-08-6
:
Formula:C₆H₆F₂N₂O₂
MDL Number:MFCD31417685
:176.121 g/mol
Storage Temperature:Ambient
Frequently Bought Together