0 zł
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Poland
ContactUsLinkComponent
Methyl 5-(difluoromethyl)pyrazine-2-carboxylate
Methyl 5-(difluoromethyl)pyrazine-2-carboxylate 95%
Methyl 5-(difluoromethyl)pyrazine-2-carboxylate 95%
Methyl 5-(difluoromethyl)pyrazine-2-carboxylate 95%
APOSPC520789-1G
:
Apollo Scientific
:
Methyl 5-(difluoromethyl)pyrazine-2-carboxylate 95%
APOSPC520789-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
188.132 g/mol
:
Ambient
:
C₇H₆F₂N₂O₂
:
MFCD30697523