2-(Difluoromethyl)-3-fluoro-6-nitro-phenol
Catalog # APOSPC520345-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:207.107 g/mol
Storage Temperature:Ambient
CAS Number:2090420-72-5
Formula:C₇H₄F₃NO₃
MDL Number:MFCD30496652