1-Chloro-4-(difluoromethyl)-5-fluoro-2-methoxy-benzene
Catalog # APOSPC520344-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:210.581 g/mol
Storage Temperature:Ambient
CAS Number:2149589-82-0
Formula:C₈H₆ClF₃O
MDL Number:MFCD29767790