1-Bromo-3-(difluoromethyl)-4-fluoro-2-methoxy-benzene
Catalog # APOSPC520343-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:255.032 g/mol
Storage Temperature:Ambient
CAS Number:2091158-36-8
Formula:C₈H₆BrF₃O
MDL Number:MFCD29767789