2-(Difluoromethyl)-3-fluoro-aniline
Catalog # APOSPC520337-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:161.124 g/mol
Storage Temperature:Ambient
CAS Number:2091605-37-5
Formula:C₇H₆F₃N
MDL Number:MFCD30496649