5-Chloro-2-(difluoromethyl)-3-fluoro-phenol
Catalog # APOSPC520333-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:196.554 g/mol
Storage Temperature:Ambient
CAS Number:2092102-01-5
Formula:C₇H₄ClF₃O
MDL Number:MFCD30496645