1-(4-Chlorophenoxy)-2-(difluoromethyl)-3-fluoro-benzene
Catalog # APOSPC520329-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Rodzaj opakowania:Bottle
- Ilość:1 g
- Seq:3047
Specifications
About this item
:
:272.65 g/mol
Storage Temperature:Temperatura otoczenia
CAS Number:2149602-13-9
Formula:C₁₃H₈ClF₃O
MDL Number:MFCD30496642