1-(Difluoromethyl)-2-fluoro-4-(4-methylsulphanylphenyl)benzene
Catalog # APOSPC520327-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:268.297 g/mol
Storage Temperature:Ambient
Formula:C₁₄H₁₁F₃S
MDL Number:MFCD30496641