Methyl 4-[4-(difluoromethyl)phenyl]benzoate
Catalog # APOSPC520304-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:262.251 g/mol
Storage Temperature:Ambient
Formula:C₁₅H₁₂F₂O₂
MDL Number:MFCD29767775