2-Bromo-4-(difluoromethyl)-6-fluoro-aniline
Catalog # APOSPC520298-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:240.021 g/mol
Storage Temperature:Ambient
CAS Number:2092183-04-3
Formula:C₇H₅BrF₃N
MDL Number:MFCD30496617