3-[4-(Difluoromethyl)-N-methyl-anilino]propanenitrile
Catalog # APOSPC520290-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:210.223 g/mol
Storage Temperature:Ambient
Formula:C₁₁H₁₂F₂N₂
MDL Number:MFCD30496610