2-cyclobutyl-5-(difluoromethyl)thiazole
Catalog # APOSPC520280-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:189.226 g/mol
Storage Temperature:Ambient
Formula:C₈H₉F₂NS
MDL Number:MFCD29767761