4-Chloro-5-(difluoromethyl)-2-(1-piperidyl)thiazole
Catalog # APOSPC520276-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:252.712 g/mol
Storage Temperature:Ambient
Formula:C₉H₁₁ClF₂N₂S
MDL Number:MFCD29767757