Order Entry
Poland
ContactUsLinkComponent
2-(4-Bromophenyl)-4-(difluoromethyl)thiazole
2-(4-Bromophenyl)-4-(difluoromethyl)thiazole
Catalog # APOSPC520272-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(4-Bromophenyl)-4-(difluoromethyl)thiazole
Catalog # APOSPC520272-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :290.127 g/mol
Storage Temperature:Ambient
CAS Number:2092628-74-3
Formula:C₁₀H₆BrF₂NS
MDL Number:MFCD29767753