Skip to Header
Skip to Main Content
Skip to Footer
Zaawansowany
Zaawansowany
SDS
Certyfikaty
Dane producenta
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Poland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-(2-Bromophenyl)-4-(difluoromethyl)thiazole
2-(2-Bromophenyl)-4-(difluoromethyl)thiazole
2-(2-Bromophenyl)-4-(difluoromethyl)thiazole
APOSPC520265-1G
:
Apollo Scientific
:
2-(2-Bromophenyl)-4-(difluoromethyl)thiazole
APOSPC520265-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
290.127 g/mol
:
Ambient
:
2091121-59-2
:
C₁₀H₆BrF₂NS
:
MFCD29767745