3-[5-(Difluoromethyl)-2-thienyl]pyridin-2-ol
Catalog # APOSPC520245-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:227.23 g/mol
Storage Temperature:Ambient
CAS Number:2091581-96-1
Formula:C₁₀H₇F₂NOS
MDL Number:MFCD29767724