Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Poland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Chloro-4-[5-(difluoromethyl)-3-thienyl]phenol
2-Chloro-4-[5-(difluoromethyl)-3-thienyl]phenol
2-Chloro-4-[5-(difluoromethyl)-3-thienyl]phenol
APOSPC520228-1G
:
Apollo Scientific
:
2-Chloro-4-[5-(difluoromethyl)-3-thienyl]phenol
APOSPC520228-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
260.687 g/mol
:
Ambient
:
C₁₁H₇ClF₂OS
:
MFCD29767708