[4-[5-(Difluoromethyl)-2-thienyl]phenyl]azinate
Catalog # APOSPC520207-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:255.241 g/mol
Storage Temperature:Ambient
Formula:C₁₁H₇F₂NO₂S
MDL Number:MFCD30496624