3-Bromo-5-(difluoromethyl)-2-propyl-thiophene
Catalog # APOSPC520206-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:255.123 g/mol
Storage Temperature:Ambient
Formula:C₈H₉BrF₂S
MDL Number:MFCD29767687