3-Bromo-5-[5-(difluoromethyl)-2-thienyl]phenol
Catalog # APOSPC520203-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:305.138 g/mol
Storage Temperature:Ambient
Formula:C₁₁H₇BrF₂OS
MDL Number:MFCD29767684