2-(Difluoromethyl)thieno[2,3-b]pyridine
Catalog # APOSPC520197-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:185.194 g/mol
Storage Temperature:Ambient
Formula:C₈H₅F₂NS
MDL Number:MFCD29767678