Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Poland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Chloro-4-(difluoromethyl)-2-ethyl-1H-imidazole
5-Chloro-4-(difluoromethyl)-2-ethyl-1H-imidazole
5-Chloro-4-(difluoromethyl)-2-ethyl-1H-imidazole
APOSPC520179-1G
:
Apollo Scientific
:
5-Chloro-4-(difluoromethyl)-2-ethyl-1H-imidazole
APOSPC520179-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
180.583 g/mol
:
Ambient
:
C₆H₇ClF₂N₂
:
MFCD29767659