2-(Difluoromethyl)-4-iodo-1H-pyrrole
Catalog # APOSPC520176-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:242.993 g/mol
Storage Temperature:Ambient
CAS Number:2091786-73-9
Formula:C₅H₄F₂IN
MDL Number:MFCD29767656