4-Chloro-5-(difluoromethyl)-7H-pyrrolo[2,3-d]pyrimidine
Catalog # APOSPC520168-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:203.577 g/mol
Storage Temperature:Ambient
CAS Number:2090261-97-3
Formula:C₇H₄ClF₂N₃
MDL Number:MFCD29767648