4-Bromo-3-(difluoromethyl)-1H-indole
Catalog # APOSPC520107-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:246.051 g/mol
Storage Temperature:Ambient
CAS Number:1780756-02-6
Formula:C₉H₆BrF₂N
MDL Number:MFCD29767590