3-(Difluoromethyl)-5-fluoro-1H-indole
Catalog # APOSPC520106-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:185.146 g/mol
Storage Temperature:Ambient
Formula:C₉H₆F₃N
MDL Number:MFCD29767589