2-(4-Bromophenyl)-3-(difluoromethyl)-1H-indole
Catalog # APOSPC520097-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:322.147 g/mol
Storage Temperature:Ambient
Formula:C₁₅H₁₀BrF₂N
MDL Number:MFCD29767580