Order Entry
Poland
ContactUsLinkComponent
3-(Difluoromethyl)-7-ethyl-1H-indole
3-(Difluoromethyl)-7-ethyl-1H-indole
Catalog # APOSPC520092-1G
Supplier:  Apollo Scientific
CAS Number:  
3-(Difluoromethyl)-7-ethyl-1H-indole
Catalog # APOSPC520092-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :195.209 g/mol
Storage Temperature:Ambient
Formula:C₁₁H₁₁F₂N
MDL Number:MFCD29767576