Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Poland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-(Difluoromethyl)-2-(4-fluorophenyl)-1H-indole
3-(Difluoromethyl)-2-(4-fluorophenyl)-1H-indole
3-(Difluoromethyl)-2-(4-fluorophenyl)-1H-indole
3-(Difluoromethyl)-2-(4-fluorophenyl)-1H-indole
APOSPC520091-1G
:
Apollo Scientific
:
3-(Difluoromethyl)-2-(4-fluorophenyl)-1H-indole
APOSPC520091-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
261.242 g/mol
:
Ambient
:
C₁₅H₁₀F₃N
:
MFCD29767575