Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Poland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
7-Bromo-3-(difluoromethyl)-1H-indole
7-Bromo-3-(difluoromethyl)-1H-indole
7-Bromo-3-(difluoromethyl)-1H-indole
APOSPC520086-1G
:
Apollo Scientific
:
7-Bromo-3-(difluoromethyl)-1H-indole
APOSPC520086-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
246.051 g/mol
:
Ambient
:
1780756-10-6
:
C₉H₆BrF₂N
:
MFCD29767570