3-(Difluoromethyl)-1,6,7,8-tetrahydrocyclopenta[g]indole
Catalog # APOSPC520081-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:207.219 g/mol
Storage Temperature:Ambient
Formula:C₁₂H₁₁F₂N
MDL Number:MFCD29767565