3-Chloro-2-(difluoromethyl)-1H-indole
Catalog # APOSPC520070-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:201.6 g/mol
Storage Temperature:Ambient
Formula:C₉H₆ClF₂N
MDL Number:MFCD29767554