2-(Difluoromethyl)-5-methyl-pyrazine
Catalog # APOSPC520069-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:144.122 g/mol
Storage Temperature:Ambient
Formula:C₆H₆F₂N₂
MDL Number:MFCD29767553