5-Bromo-3-(difluoromethyl)pyrazin-2-amine
Catalog # APOSPC520068-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:224.006 g/mol
Storage Temperature:Ambient
CAS Number:2092522-38-6
Formula:C₅H₄BrF₂N₃
MDL Number:MFCD29767552