Methyl 2-(difluoromethyl)pyrimidine-5-carboxylate
Catalog # APOSPC520065-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:188.132 g/mol
Storage Temperature:Ambient
CAS Number:2120555-73-7
Formula:C₇H₆F₂N₂O₂
MDL Number:MFCD29767548