Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0 zł
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Poland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-(Difluoromethyl)-2-methoxy-pyrimidine
5-(Difluoromethyl)-2-methoxy-pyrimidine
5-(Difluoromethyl)-2-methoxy-pyrimidine
APOSPC520058-1G
:
Apollo Scientific
:
5-(Difluoromethyl)-2-methoxy-pyrimidine
APOSPC520058-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
160.121 g/mol
:
Ambient
:
2128733-64-0
:
C₆H₆F₂N₂O
:
MFCD29767541