5-(Difluoromethyl)-2-methylsulphanyl-pyrimidin-4-amine
Catalog # APOSPC520050-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:191.202 g/mol
Storage Temperature:Ambient
CAS Number:2090187-83-8
Formula:C₆H₇F₂N₃S
MDL Number:MFCD29767533