Order Entry
Poland
ContactUsLinkComponent
5-(Difluoromethyl)-2-isopropyl-pyrimidine
5-(Difluoromethyl)-2-isopropyl-pyrimidine
Catalog # APOSPC520043-1G
Supplier:  Apollo Scientific
CAS Number:  
5-(Difluoromethyl)-2-isopropyl-pyrimidine
Catalog # APOSPC520043-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :172.175 g/mol
Storage Temperature:Ambient
Formula:C₈H₁₀F₂N₂
MDL Number:MFCD29767526