2-Chloro-3-(difluoromethyl)-8-methylquinoline
Catalog # APOSPC520037-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:227.638 g/mol
Storage Temperature:Ambient
CAS Number:2090445-82-0
Formula:C₁₁H₈ClF₂N
MDL Number:MFCD29767521