2-Chloro-3-(difluoromethyl)-7-methoxyquinoline
Catalog # APOSPC520032-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:243.637 g/mol
Storage Temperature:Ambient
CAS Number:2091217-12-6
Formula:C₁₁H₈ClF₂NO
MDL Number:MFCD29767516