1H,1H-Hexafluorobut-1-ene (HFO-1336mcyf)
Catalog # APOSPC51055-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Cylinder
- Pk:5 g
- Seq:3173
- Pk Info:1/4" NPTM connection
Specifications
About this item
:
:164.049 g/mol
Storage Temperature:Ambient
CAS Number:374-39-0
Formula:C₄H₂F₆
MDL Number:MFCD18074334