Order Entry
Poland
ContactUsLinkComponent
Ethyl 4-(2-Ethoxy-1,1-difluoro-2-oxoethyl)benzoate
Ethyl 4-(2-Ethoxy-1,1-difluoro-2-oxoethyl)benzoate
Catalog # APOSPC510333-5G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl 4-(2-Ethoxy-1,1-difluoro-2-oxoethyl)benzoate
Catalog # APOSPC510333-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :272.245 g/mol
Storage Temperature:Ambient
CAS Number:924312-34-5
Formula:C₁₃H₁₄F₂O₄
MDL Number:MFCD29065894