Order Entry
Poland
ContactUsLinkComponent
Ethyl 2-(2-Chlorophenyl)-2,2-difluoroacetate
Ethyl 2-(2-Chlorophenyl)-2,2-difluoroacetate
Catalog # APOSPC510325-5G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl 2-(2-Chlorophenyl)-2,2-difluoroacetate
Catalog # APOSPC510325-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :234.627 g/mol
Storage Temperature:Ambient
CAS Number:1248075-48-0
Formula:C₁₀H₉ClF₂O₂
MDL Number:MFCD16748873