Złóż zamówienie
Poland
ContactUsLinkComponent
Ethyl 2-(3-Cyanophenyl)-2,2-difluoroacetate
Ethyl 2-(3-Cyanophenyl)-2,2-difluoroacetate
Catalog # APOSPC510321-1G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl 2-(3-Cyanophenyl)-2,2-difluoroacetate
Catalog # APOSPC510321-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :225.191 g/mol
Storage Temperature:Ambient
CAS Number:1249788-93-9
Formula:C₁₁H₉F₂NO₂
MDL Number:MFCD16756772