Order Entry
Poland
ContactUsLinkComponent
1,3-Dibromo-2-(difluoromethoxy)benzene
1,3-Dibromo-2-(difluoromethoxy)benzene
Catalog # APOSPC510285-1G
Supplier:  Apollo Scientific
CAS Number:  
1,3-Dibromo-2-(difluoromethoxy)benzene
Catalog # APOSPC510285-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :301.911 g/mol
Storage Temperature:Ambient
CAS Number:1182728-50-2
Formula:C₇H₄Br₂F₂O
MDL Number:MFCD22418262