Order Entry
Poland
ContactUsLinkComponent
2-[2-Fluoro-4-(methylsulphonyl)phenyl]-1,3-dioxolane
2-[2-Fluoro-4-(methylsulphonyl)phenyl]-1,3-dioxolane
Catalog # APOSPC510100-5G
Supplier:  Apollo Scientific
CAS Number:  
2-[2-Fluoro-4-(methylsulphonyl)phenyl]-1,3-dioxolane
Catalog # APOSPC510100-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :246.255 g/mol
Storage Temperature:Ambient
CAS Number:1346597-53-2
Formula:C₁₀H₁₁FO₄S
MDL Number:MFCD20491403