2-Chloro-4-(difluoromethyl)-1-iodobenzene 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1261821-31-1
:
Formula:C₇H₄ClF₂I
MDL Number:MFCD18392844
:288.461 g/mol
Storage Temperature:Ambient
UN:3265
ADR:8,II
Frequently Bought Together