
5-Chloro-6-(difluoromethyl)pyrimidin-4-ol
Catalog # APOSPC50327-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Rodzaj opakowania:Bottle
- Ilość:100 mg
- Seq:2440
Specifications
About this item
:
:180.54 g/mol
:GHS07
Storage Temperature:Temperatura otoczenia
CAS Number:425394-28-1
Formula:C₅H₃ClF₂N₂O
MDL Number:MFCD09909765