2,6-Difluoro-4-(trifluoromethyl)phenyl 4-fluorophenyl ether 95%
Catalog # APOSPC502832-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:292.177 g/mol
Storage Temperature:Ambient
CAS Number:2244084-61-3
Formula:C₁₃H₆F₆O
MDL Number:MFCD31629971